Learn More
3B Scientific™ Cardionics SAM 3G™ Student Auscultation Manikin
3B Scientific™ Cardionics SAM 3G™ Student Auscultation Manikin is the world's most innovative auscultation product currently on the market
Supplier: 3B Scientific™ 1021554
Description
Product Features:
- Auscultation is an essential clinical skill needed to assess and monitor patients conditions
- The new Cardionics SAM 3G Student Auscultation Manikin 3rd Generation makes teaching and learning this skill even more versatile and economic
- Can be used with any stethoscope available and offers the largest Real Sound Library, Case Videos, Echo Cardiogram Videos and ECG Waveforms for students to master identification of various heart, lung and bowel sounds
- All sounds and videos are recorded from live patients for students to experience a life-like simulation
- Case videos contain patient and clinical interactions for various conditions and enhance the learning by providing visual representation of the sounds
- When connected to the laptop (Included) with the pre-installed software, a variety of sounds, videos and lessons recorded can be accessed
- When connected to speakers (Sold Separately) SAM 3G Student Auscultation Manikin can be easily moved into a classroom or an auditorium for group instruction
- The programmable and password protected lectures allow teaching institutions to have numerous instructors utilize the SAM 3G Trainer
Real Sound Library for Auscultation Training includes:
- Normal Sensi 75 BPM
- Physiological Splitting of S2 UM 70 BPM
- Aortic Septal Defect Sensi 80 BPM
- Central Shunt UM 150 BPM
- Innocent Murmur Sensi 85 BPM
- Mitral Regurgitation Sensi 95 BPM
- MVP Sensi 90 BPM
- Patent Ductus Arteriosus UM 96 BPM
- Patent Ductus Arteriosus Sensi 95 BPM
- Pulmonary Stenosis and Patent Ductus Arteriosus UM 138 BPM
- Pulmonary Stenosis Sensi 88 BPM
- Restrictive Ventricular Septal Defect UM 72 BPM
- Tetralogy of Fallot Sensi 120 BPM
- Ventricular Septal Defect Sensi 76 BPM
- Ventricular Septal Defect UM 130 BPM
- Bronchial Breath 13 BPM
- BronchoVesicular 26 BPM
- Vesicular Normal 16 BPM
- Fine Crackles 10 BPM
- Late Inspiratory Crackles 14 BPM
- Low Pitched Crackles 24 BPM
- Pleural Friction Rub 28 BPM
- Velcro Crackles 10 BPM
- Wheeze High Pitched 16 BPM
- Wheeze Low Pitched 34 BPM
How is SAM 3G Student Auscultation Manikin different?
- It is lightweight & portable
- It has a new and user friendly software program
- Phonocardiogram
- It has the largest library of sounds (24 sounds) & videos (36 videos altogether)
- It has the complete Lesson Guide
- It can be used with any stethoscope
- It has a palpable carotid pulse, ECG videos and waveform for heart sounds
- Users can create and save own case videos
- Users can setup password protected lectures for student assessment
Specifications
Manikin | |
Light Skin | |
41.89 lb. |
Mastering Identification of Various Heart, Lung and Bowel Sounds | |
SAM 3G Male Auscultation Torso, DELL Laptop Computer, Real Sound Library, Echo Cardiogram Videos, Case Videos, SAM's Lesson Guide, SAM T-Shirt, User Manual |