Learn More
3B Scientific™ Cardionics SAM Basic™ Student Auscultation Manikin
3B Scientific™ Cardionics SAM Basic™ Student Auscultation Manikin with WiFi for teaching or learning heart, lung, and bowel sounds
Specifications
Product Type | Manikin |
---|---|
For Use With (Application) | Teaching or Learning Heart, Lung, and Bowel Sounds |
Includes | SAM Basic Male Auscultation Torso, SimScope Stethoscope, SimScope Software, Site License, USB Cable, SAM T-Shirt, User manual |
Weight (English) | 14.55 lb. |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
S27948
|
3B Scientific™
1020099 |
Light Skin | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
S27949
|
3B Scientific™
1022475 |
Dark Skin | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Product Features:
- Auscultation is an essential clinical skill needed to assess and monitor patients' conditions
- SAM Basic™ Trainer can be used with the SimScope stethoscope (included)
- All sounds and videos are recorded from adult live patients for students to experience a life-like simulation
- SimScope stethoscope is used to simulate adult heart, lung and bowel sounds, utilizing a large sound library incorporated into the SimScope stethoscope
- WiFi upgrade enables wireless communication between the SimScope and the SAM Basic manikin
- Users are able to change sounds remotely or in conjunction with a scenario-based simulation
- By placing the SimScope over the anatomically correct locations of SAM Basic, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills.
- Uses include: individual learning, group lectures, and for viewing phonocardiograms
Delivery content:
- A portable SAM Basic Male Auscultation Torso
- One SimScope stethoscope
- The SimScope software and site license
- USB cable
- One SAM t-shirt
- User's manual
How is SAM Basic Student Auscultation Manikin different?
- It is lightweight & portable
- It utilizes the extensive Cardionics proprietary heart, lung, bowel and bruit sounds library
- Students can listen at anatomically correct auscultation sites
- Customized Library is available to fit specific scenarios and programs
- The SimScope WiFi upgrade allows for remote selection of sound scenarios
Specifications
Manikin | |
Teaching or Learning Heart, Lung, and Bowel Sounds | |
SAM Basic Male Auscultation Torso, SimScope Stethoscope, SimScope Software, Site License, USB Cable, SAM T-Shirt, User manual | |
14.55 lb. |