Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardionogen 1, Tocris Bioscience™

Catalog Number 447510
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
577696-37-8
C13H14N4OS
274.342
SWTUPSNBTSPBIH-UHFFFAOYSA-N
cardionogen 1, 6-cyclohexyl-3-furan-2-yl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-2-furanyl-1,2,4-triazolo 3,4-b 1,3,4 thiadiazole, cardionogen hplc, 6-cyclohexyl-3-2-furanyl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-2-furyl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-furan-2-yl 1,2,4 triazolo 3,4-b 1,3,4 thiadiazole
663145
6-cyclohexyl-3-(furan-2-yl)-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazole
C1CCC(CC1)C2=NN3C(=NN=C3S2)C4=CC=CO4
This item is not returnable. View return policy
577696-37-8
C13H14N4OS
274.342
SWTUPSNBTSPBIH-UHFFFAOYSA-N
cardionogen 1, 6-cyclohexyl-3-furan-2-yl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-2-furanyl-1,2,4-triazolo 3,4-b 1,3,4 thiadiazole, cardionogen hplc, 6-cyclohexyl-3-2-furanyl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-2-furyl-1,2,4 triazolo 3,4-b 1,3,4 thiadiazole, 6-cyclohexyl-3-furan-2-yl 1,2,4 triazolo 3,4-b 1,3,4 thiadiazole
663145
6-cyclohexyl-3-(furan-2-yl)-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazole
C1CCC(CC1)C2=NN3C(=NN=C3S2)C4=CC=CO4

Inhibitor of Wnt/β-catenin signaling

Wnt signaling modulator. Potently inhibits Wnt/β-catenin-dependent transcription in murine ES cells (EC50 = 23 nM) and zebrafish embryos. Enlarges heart size via cardiomyocyte hyperplasia; induces ES cell cardiac differentiation.
Quantity 10 mg
Formula Weight 274.34
Percent Purity >99%
Chemical Name or Material Cardionogen 1
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit