Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardiotrophin-1/CT-1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17947420UL

 View more versions of this product

Catalog No. NBP17947420


Add to cart

Description

Description

Cardiotrophin-1/CT-1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Cardiotrophin-1/CT-1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
CTF1
Synthetic peptide directed towards the N terminal of human CTF1. Peptide sequence HSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAG.
21 kDa
20ul
Cell Cycle and Replication, Protein Kinase
Primary
1489
Western Blot
Western Blot 1:1000
NP_001321
cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit