Learn More
Cardiotrophin-1/CT-1 Rat anti-Mouse, Alexa Fluor 488, Clone: RM0155-1J24, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212067AF488
Catalog No. NB212067488
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Cardiotrophin-1/CT-1 Monoclonal antibody specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Cardiotrophin-1/CT-1
RM0155-1J24
Western Blot
Purified
CTF1
Mouse Cardiotrophin 1
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Protein Kinase
Primary
1489
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse