Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardiotrophin-1/CT-1 Rat anti-Mouse, Clone: RM0155-1J24, Novus Biologicals™
Rat Monoclonal Antibody
Specifications
Antigen | Cardiotrophin-1/CT-1 |
---|---|
Clone | RM0155-1J24 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212067
|
Novus Biologicals
NBP212067 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019026
|
Novus Biologicals
NBP2120670.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Cardiotrophin-1/CT-1 Monoclonal antibody specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot.Specifications
Cardiotrophin-1/CT-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Cell Cycle and Replication, Protein Kinase | |
Q60753 | |
1489 | |
Mouse Cardiotrophin 1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0155-1J24 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1 | |
CTF1 | |
IgG2 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title