Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiotrophin 1 (CTF1) Mouse ELISA Kit Non-distribution product as customer accommodation.

Sandwich ELISA Kit

$491.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 200 pg/mL
Assay Range 0.246-60 ng/mL
Test Time 1 hr. 20 min.
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EMCTF1 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
EMCTF1
96 Tests
Pack for $491.00
Add to cart
 
Description

Description

The Mouse Cardiotrophin 1 (CT-1) ELISA quantitates Ms CT-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CT-1. Principle of the method The Mouse CT-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cardiotrophin-1 (CT-1) is a member of the IL-6 family of cytokines, which signal through gp130 receptor complexes. gp130 complexes with several different receptor subunits to transmit signals from Cardiotrophin-1, IL-6, LIF, OSM, CNTF and IL-11. Cardiotrophin-1 binds to and activates the leukemia inhibitory factor (LIF) receptor/gp130 receptor complex and has been shown to induce hypertrophy in cardiac myocytes in vitro. Cardiotrophin-1, a secreted protein expressed at high levels in myocardium during cardiogenesis, has been shown to promote proliferation and survival of embryonic cardiomyocytes, suggesting a role for Cardiotrophin-1 in the activation of gp130 during cardiac development.
Specifications

Specifications

2°C to 8°C
0.246-60 ng/mL
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
<10%
200 pg/mL
1 hr. 20 min.
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit