Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARF Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549978

Catalog No. PA549978


Add to cart

Description

Description

The antibody detects endogenous levels of total CARF / ALS2CR8 protein.

Acts as a transcriptional activator that mediates the calcium- and neuron-selective induction of BDNF exon III transcription. Binds to the consensus calcium-response element CaRE1 5 ft.-CTATTTCGAG-3 ft. sequence. Hadano S., Nat. Genet. 29:166-173(2001). Tao X., Neuron 33:383-395(2002). Hillier L.W., Nature 434:724-731(2005)
Specifications

Specifications

CARF
Polyclonal
Unconjugated
CARF
Liquid
CARF
Synthesized peptide derived from internal of human ALS2CR8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8N187
ALS2CR8
Rabbit
IgG
100 μL
-20°C
Primary
79800
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit