Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cargille Immersion Oil
SCI_ED

Finest immersion oil available.

$21.50 - $42.75

Specifications

Certifications/Compliance ISO 8036/1
Refractive Index 1.5150 ±0.0002
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Viscosity Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Viscosity Type Price Quantity & Availability  
S19312
SCI_ED
View Documents Promotion Details
Cargille
16484/4OZ/EMD
4 oz. 1250 cS Type B Immersion Oil
Each for $42.75
Case of 12 EA for $511.00
Add to cart
 
S19304A
SCI_ED
View Documents Promotion Details
Cargille
16482/1OZ/EMD
1 oz. 150 cS Type A Immersion Oil
Each for $21.50
Add to cart
 
S19305
SCI_ED
View Documents Promotion Details
Cargille
16482/4OZ/EMD
4 oz. 150 cSt Type A Immersion Oil
Each for $42.75
Case of 12 EA for $511.00
Add to cart
 
S19314
SCI_ED
View Documents Promotion Details
Cargille
16484/1OZ/EMD
1 oz. 1250 cSt Type B Immersion Oil This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

For use with 100X oil immersion objectives. Type B is recommended for most educational microscopy; contains no PCB compounds.

  • Type A: Low fluorescence, low viscosity
  • Type B: Low fluorescence, high viscosity

Specifications

Specifications

ISO 8036/1
1.5150 ±0.0002
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit