Learn More
Carhartt™ Denim Pant
Manufacturer:  CARHARTT INC FRB13DNMW40WX32L
 View more versions of this product
Catalog No. 18-799-818
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
15.2 Calories/cm2
NFPA 70E™, NFPA 2112
5.0
40 x 32 in.
19 in.
Denim
Arc-resistant high-tension snap closure, Brass zipper
Cotton
Male
48.26cm