Learn More
Carhartt™ Denim Pant
Manufacturer:  CARHARTT INC FRB13DNMW36WX30L
 View more versions of this product
Catalog No. 18-799-813
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
15.2 Calories/cm2
NFPA 70E™, NFPA 2112
5.0
36 x 30 in.
19 in.
Denim
Arc-resistant high-tension snap closure, Brass zipper
Cotton
Male
48.26cm