Carhartt™ Flame-Resistant Relaxed Fit Denim Jeans
Manufacturer: CARHARTT INC FRB160DNMB54X32
 View more versions of this product
Catalog No. 18-799-452
  Check Availability
Add to Cart
    Specifications
    Certifications NFPA 70E and NFPA 2112
    Product Type Flame-Resistant Relaxed Fit Denim Jean
    Material Denim
    Size 54 x 32 Big/Tall
    Arc Flash Rating ATPV 25 calories/cm2
  • Description & Specifications
Related Products
Product Content Feedback