Learn More
Carhartt™ Flame-Resistant Relaxed Fit Denim Jeans
Manufacturer:  CARHARTT INC FRB160DNM3630
 View more versions of this product
Catalog No. 18-799-836
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 70E and NFPA 2112
ATPV 25 calories/cm2
Denim
Flame-Resistant Relaxed Fit Denim Jean
36 x 30