Learn More
Carhartt™ Flame-Resistant Relaxed Fit Denim Jeans
Manufacturer:  CARHARTT INC FRB160DNM34X32
 View more versions of this product
Catalog No. 18-799-372
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 70E and NFPA 2112
ATPV 25 calories/cm2
34 x 32 Regular
Flame-Resistant Relaxed Fit Denim Jean
Denim