Learn More
Carhartt™ Ladies Pant
Manufacturer:  CARHARTT INC WFRB159DNY18X32
 View more versions of this product
Catalog No. 18-799-831
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ATPV 12.1 Calories/cm2
NFPA 70E™, NFPA 2112
Cotton, High-tenacity Nylon
Cotton, Nylon
18 x 32 in.
Dark Navy
Arc-resistant high-tension snap closure at waist
2.0
Triple stitched
Female