Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carhartt Men's Duck Active Thermal Lined Jackets

Manufacturer:  CARHARTT INC J131BRNREGMED

 View more versions of this product

Catalog No. 18-999-137


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Zipper
Medium
Ring-spun Cotton Duck
64.77cm
Polyester Thermal
38 to 40 in.
Brown
Thermal-lined hood with drawcord
25.5 in.
Triple stitched
4.0
Regular
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit