Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carhartt Men's Duck Active Thermal Lined Jackets

Manufacturer:  CARHARTT INC J131BRNTALL2XL

 View more versions of this product

Catalog No. 18-999-138


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Zipper
2X-Large
Ring-spun Cotton Duck
72.39cm
Polyester Thermal
50 to 52 in.
Brown
Thermal-lined hood with drawcord
28.5 in.
Triple stitched
4.0
Tall
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit