Learn More
Carhartt™ Men's Flame-Resistant Twill Shirts with Pocket Flap
Manufacturer:  CARHARTT INC FRS160DNYM
 View more versions of this product
Catalog No. 18-799-332
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ATPV 8.6 Calories/cm2
Button-down
Dark Navy
Medium
Button-down collar
Meets NFPA 2112 and 70E compliant
Button
7 oz. Flame resistant twill
Male
2 Chest pockets with flaps, button closures, FR melamine buttons, button-down collar, triple-stitched main seams