Carhartt™ Men's Flame-Resistant Twill Shirts with Pocket Flap
Manufacturer: CARHARTT INC FRS160DNYMEDREG
 View more versions of this product
Catalog No. 18-799-411
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Arc Flash Rating ATPV 8.6 calories/cm2
    Material 7 oz. Twill
    Size Medium Regular
    Color Dark Navy
  • Description & Specifications
Related Products
Product Content Feedback