Carhartt™ Men's Flame-Resistant Twill Shirts with Pocket Flap
Manufacturer: CARHARTT INC FRS160MBL3XLTLL
 View more versions of this product
Catalog No. 18-799-424
  Check Availability
Add to Cart
    Specifications
    Arc Flash Rating ATPV 8.6 calories/cm2
    Material 7 oz. Twill
    Size 3X-Large Big/Tall
    Color Medium Blue
  • Description & Specifications
Related Products
Product Content Feedback