Carhartt™ Navy Jean
Manufacturer: CARHARTT INC FRB159DNY32X34
 View more versions of this product
Catalog No. 18-799-511
  Check Availability
Add to Cart
    Specifications
    No. of Pockets 2.0
    Material Cotton, Nylon
    Seam Type Triple stitched
    Size 32 x 34 in.
    Arc Flash Rating ATPV 12.1 Calories/cm2
    Color Dark Navy
    Certifications/Compliance NFPA 70E™, NFPA 2112
    Closure Type Arc-resistant high-tension snap closure at waist
    Construction Cotton, High-tenacity Nylon
  • Description & Specifications
Related Products
Product Content Feedback