Learn More
Pyramex™ Carhartt™ Toccoa™ Safety Eyewear
Click to view available options
Frame Color:
Black, Tan
Realtree Xtra™ Camouflage
Lens Tint:
Bronze
Clear
Gray
Description
Specifications
Lens Type | H2MAX |
Certifications/Compliance | ANSI Z87.1+,CSA Z94.3 |
Lens Tint | Gray |
Frame Color | Realtree Xtra™ Camouflage |
Weight (Metric) | 42 g |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |