Carhartt™ Women's Flame-Resistant Relaxed Fit Denim Jeans
Manufacturer: CARHARTT INC WFRB160DNM18X30
 View more versions of this product
Catalog No. 18-799-439
  Check Availability
Add to Cart
    Specifications
    Material 14.75 oz. Denim
    Size 18 x 30 Relaxed Fit
    Arc Flash Rating ATPV 25 calories/cm2
  • Description & Specifications
Related Products
Product Content Feedback