Carhartt™ Work-Dry™ Men's Flame-Resistant Long-Sleeve T-Shirts
Manufacturer: CARHARTT INC FRK009DNYM
 View more versions of this product
Catalog No. 18-799-930
  Check Availability
Add to Cart
    Specifications
    Material 5.5 oz. 48% Modacrylic/48% Tencel/4% Spandex
    Size Medium Regular
    Sleeve Style Long-Sleeve
    Gender Male
    Resistance Fire
    Color Dark Navy
  • Description & Specifications
Related Products
Product Content Feedback