Carhartt™ Work-Dry™ Men's Flame-Resistant Short-Sleeve T-Shirts
Manufacturer: CARHARTT INC FRK008SNDTALL3X
 View more versions of this product
Catalog No. 18-799-921
  Check Availability
Add to Cart
    Specifications
    Material 5.5 oz. 48% Modacrylic/48% Tencel/4% Spandex
    Size 3X-Large Tall
    Color Sand
  • Description & Specifications
Related Products
Product Content Feedback