Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARHSP1 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP300026AF647
Description
CARHSP1 Polyclonal antibody specifically detects CARHSP1 in Human samples. It is validated for Western Blot, ELISASpecifications
CARHSP1 | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
calcium regulated heat stable protein 1 (24kD), calcium regulated heat stable protein 1, 24kDa, calcium-regulated heat stable protein 1, calcium-regulated heat-stable protein (24kD), Calcium-regulated heat-stable protein of 24 kDa, CRHSP-24MGC111446, CSDC1 | |
Produced in rabbits immunized with purified, recombinant CARHSP1 (Q9Y2V2; Met1-Ser147). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
ELISA, Western Blot | |
Western Blot, ELISA | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Signal Transduction | |
Primary | |
23589 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title