Learn More
Abnova™ CARKL 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00023729T01
Catalog No. 89-938-039
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CARKL 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 52.69kDa
Transient overexpression cell lysate was tested with anti-CARKL antibody (Mfr. No. H00023729-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_013276.2
pCMV-CARKL full-length
100 μL
Human