AxioLab™ A1 Microscopes
Ergonomic, easy-to-use, and durable
Manufacturer: Carl Zeiss, Inc. 4109990000017
 View more versions of this product
Catalog No. 12-070-204
  Check Availability
Add to Cart
    Specifications
    Axis Type 10X/20mm
    Design Type Right-handed mechanical
    Condenser Type 0.9/1.25 H
    Magnification Power 5x, 10x, 20x, 40x, 100x oil A-Plan
    Objective Types 5x, 10x, 20x, 40x, 100x oil A-Plan
    Eyepiece 10X, 20mm E-PL
    For Use With (Application) Bio medical or industrial laboratories, for universities and training courses
    Head Style Binocular phototube
    Lighting Type 12V/35w Halogen
    Nosepiece 5-position revolving
    Includes Phototube, Halogen bulb, stand, head, eyepieces, stage with specimen holder, nosepiece, objectives, condenser, illuminator, filter, dust cover, power supply and cord
  • Description & Specifications
Related Products
Product Content Feedback