AxioLab™ A1 Microscopes
Ergonomic, easy-to-use, and durable
Manufacturer: Carl Zeiss, Inc. 4109990000021
 View more versions of this product
Catalog No. 12-070-208
  Check Availability
Add to Cart
    Specifications
    Axis Type 10X/20mm
    Design Type Right-handed mechanical
    Condenser Type H, D, Ph1, Ph2, Ph3
    Magnification Power 5x, 10x Ph1, 40x Ph2, 100x Ph3 oil
    Objective Types 5x, 10x Ph1, 40x Ph2, 100x Ph3 oil
    Eyepiece 10X, 20mm E-PL
    For Use With (Application) Bio medical or industrial laboratories, for universities and training courses
    Head Style Binocular phototube
    Lighting Type 12V/35w Halogen
    Nosepiece 5-position revolving
    Includes Phototube, stand, head, eyepieces, stage with specimen holder, nosepiece, objectives, condenser, illuminator, filter, dust cover, power supply and cord
  • Description & Specifications
Related Products
Product Content Feedback