Learn More
Immersol™ Immersion Oil
Increases the resolving power of a microscope. Zeiss Immersol™ 518 F, Immersion Oil is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
Manufacturer:  Carl Zeiss, Inc. 0000001111808
 View more versions of this product
Catalog No. 12-070-395
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
ISO 8036-1
1.518
250mL
Immersol 518 N
Bottle