Immersol™ Immersion Oil
For Carl Zeiss Microscopes
Manufacturer: Carl Zeiss, Inc. 0000001111807
 View more versions of this product
Catalog No. 12-070-397
  Check Availability
Add to Cart
    Specifications
    Product Type Immersol 518 N
    Package Bottle
    Certifications/Compliance ISO 8036-1
    Refractive Index 1.518
    Quantity 100mL
  • Description & Specifications
Related Products
Product Content Feedback