Learn More
Primo Star™ HAL/LED Microscopes: Fixed Köhler
Robust and durable enough for repeated assembling and disassembling, Primo Star offers a complete configuration with fixed- Köhler stand, excellent optics and reasonable pricing
Manufacturer:  Carl Zeiss, Inc. 4155000051000
 View more versions of this product
Catalog No. 12-171-480
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Dust cover, oiler with 5mL immersion oil, color filter set (blue, green, yellow), 6V 30w Halogen lamp
Right-handed mechanical, with Eyepiece pointer
40x to 1000x
Color Graphic Display
Education
4-position reversed
18mm Field of View
Fixed Kohler
Abbe, 0.9/1.25
4x, 10x, 40x and 100x oil plan achromatic
10X, 18mm widefield
6V, 30w Halogen, 6V, 3w LED
Color corrected infinity