Learn More
Primo Vert™ Inverted Microscope
The new standard for quick assessment and inspection of living cells
Manufacturer:  Carl Zeiss, Inc. 4109990000004
 View more versions of this product
Catalog No. 12-070-469
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
0.4 condenser with binocular phototube
4x Ph0, 10x Ph1, LD 20x Ph1, LD 40x Ph2
20mm field of view
4-position
4x Ph0, 10x Ph1, LD 20x Ph1, LD 40x Ph2
Routine and cutting edge research lab applications such as live cell inspection, cancer and HIV research, human, animal and plant genetics, cell biology
30w Halogen
Infinity