Primo Vert™ Inverted Microscope
The new standard for quick assessment and inspection of living cells
Manufacturer: Carl Zeiss, Inc. 4109990000001
 View more versions of this product
Catalog No. 12-070-466
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product.
Please contact customer service for assistance: 1-800-766-7000.
Add to Cart
    Specifications
    Condenser Type 0.3 condenser with binocular tube
    Magnification Power 4x, 10x Ph1
    Objective Types 4x, 10x Ph1
    For Use With (Application) Routine and cutting edge research lab applications such as live cell inspection, cancer and HIV research, human, animal and plant genetics, cell biology
    Eyepiece 20mm field of view
    Lighting Type 30w Halogen
    Nosepiece 4-position
    Optical Design Infinity
  • Description & Specifications
Related Products
Product Content Feedback