Replacement Bulbs for Microscopes and Fiber Optic Power Supplies
For Carl Zeiss Microscopes and Fiber Optic Power Supplies
Manufacturer: Carl Zeiss, Inc. 381619
 View more versions of this product
Catalog No. 12-070-258
  Check Availability
Add to Cart
Alert
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
    Specifications
    Product Type Mercury burner HBO
    Voltage 110V
    Wattage 50w
    For Use With (Equipment) Axiostar Plus Student, Axiostar Plus ICS
  • Description & Specifications
Related Products
Product Content Feedback