Learn More
Replacement Bulbs for Microscopes and Fiber Optic Power Supplies
For Carl Zeiss Microscopes and Fiber Optic Power Supplies
Manufacturer:  Carl Zeiss, Inc. 386108
 View more versions of this product
Catalog No. 12-070-184
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications
Description
  • Accessory for use with Carl Zeiss Stereomicrscopes
  • Halogen bulb
Description & Specifications
Specifications
Stemi 2000/2000C, Stemi 1000 (halogen illuminator)
1.5 in.
38.1mm
10w
Halogen bulb
300 hr.
6V