Stemi™ 2000 Stereomicroscopes
For stereoscopic observation only
Manufacturer: Carl Zeiss, Inc. 4950051301000
 View more versions of this product
Catalog No. 12-070-450
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Stemi 2000 Kit with cold-light source KL 300 LED
    Voltage 9V DC
    Applications For stereoscopic observation
    Magnification Power 6.5x to 50x
    Zoom Range 7.7:1
  • Description & Specifications
Related Products
Product Content Feedback