Carmine (Certified Biological Stain), Fisher Chemical

For use in histology and cytology

$239.64

Chemical Identifiers

Chemical Name or Material Carmine
CAS [1390-65-4]
Purity Grade Certified Biological Stain
Molecular Formula C9H6F2O2
MFCD Number MFCD00167027
Synonym Alum Lake, Carminic Acid
View More Specs

  • Products 1
  • Description & Specifications
  • Safety and Handling
Products
Catalog Number Description Price Quantity Availability  

C579-25

SDS Accessories
Glass Bottle; 25g
Each for $239.64
Case of 6 EA for $1,078.37
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products