Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carmine Reagent, Ricca Chemical
For Boron determination
$51.75 - $150.00
Specifications
Chemical Name or Material | Carmine Reagent |
---|---|
Name Note | Carmine Reagent |
CAS | 1390-65-4, 7664-93-9 |
CAS Min % | 98.0, 0.05 |
CAS Max % | 100.0, 0.05 |
Specifications
Carmine Reagent | |
1390-65-4, 7664-93-9 | |
100.0, 0.05 |
Carmine Reagent | |
98.0, 0.05 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title