Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carminic acid, 70-90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1260-17-9
C22H20O13
492.389
MFCD00167028
DGQLVPJVXFOQEV-JNVSTXMASA-N
C.I. 75470,Natural Red 4
10255083
CHEBI:78310
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
CC1=C2C(=CC(=C1C(=O)O)O)C(=O)C3=C(C2=O)C(=C(C(=C3O)O)C4C(C(C(C(O4)CO)O)O)O)O
1260-17-9
C22H20O13
492.389
MFCD00167028
DGQLVPJVXFOQEV-JNVSTXMASA-N
C.I. 75470,Natural Red 4
10255083
CHEBI:78310
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
CC1=C2C(=CC(=C1C(=O)O)O)C(=O)C3=C(C2=O)C(=C(C(=C3O)O)C4C(C(C(C(O4)CO)O)O)O)O
Assay Percent Range 70-90%
Beilstein 10, II, 776
Merck Index 15, 1843
Infrared Spectrum Authentic
Formula Weight 492.39
Physical Form Crystalline Powder
Percent Purity ≥70%
Packaging Glass bottle
Color Red
Quantity 1g
Chemical Name or Material Carminic acid, 70-90%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-023-3

TSCA : TSCA