Carolina 6 in. Composite Waterproof Broad Toe Womens Boots

Manufacturer: Carolina Shoe Company CA1620W065

 View more versions of this product

Catalog No. 19-089-0037

  Check Availability
Add to Cart

    Specifications

    Gender Female
    Height (English) 6 in.
    Shank Material Nylon
    Sole Features EVA midsole, slip resistant Rubber outsole, Pillow Cushion™ insole
    Toe Style Composite
    Waterproof Yes
    Color Copper Brown
    Material Crazy Horse Leather
    Size 6.5W
    Eyelets Material Brass
    Footbed Material AG7™ Polyurethane
    Tread Style Aggressive

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit