Learn More
Carolina™ Batrachospermum, Living Products for Science Education
A freshwater red algae that has long, branched, threadlike filaments bearing dense whorl of branchlets, resembling beads on a string.
Manufacturer:  Carolina Biological SupplySupplier Diversity Partner 153515
Catalog No. S21874ND
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Description
This culture requires low light level: 50 to 100 ft candles of fluorescent light or fluorescent light from the ceiling. The medium is soil-Water and the optimum growth temperature is 22° Celsius.
  • Enough material for a class of 30 students
Specifications
Specifications
Education
1 Tube
Soil-Water
22°C
22°C
Class of 30 students
Batrachospermum
low light level: 50 to 100 ft candles of fluorescent light or fluorescent light from the ceiling