Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Cold Water Seaweed
SCI_ED

A generous portion of marine algae, freshly collected and shipped from Maine, is supplied in each shipment.

$33.75 - $49.25

Specifications

Quantity Each
For Use With (Application) For classroom
Media Type Alga-Gro™ medium
Storage Requirements 22°C
Sufficient For Class of 30 students
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 2
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
S07250ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
151420
Laminaria Laminaria
Each for $49.25
Add to cart
 
S07247ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
151430
Ulva Ulva
Each for $33.75
Add to cart
 
Description

Description

  • Each culture contains enough material for a class of 30 students

† Due to State restrictions Ulva will not ship into: WI.

Specifications

Specifications

Each
Alga-Gro™ medium
Class of 30 students
For classroom
22°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit