Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Euglenoids
SCI_ED

Euglenoids are one of the best-known groups of flagellates, commonly found in freshwater especially when it is rich in organic materials, with a few marine and endosymbiotic members.

Specifications

Quantity Each
Lighting Type Low
Media Type Soil Water and Pea
Storage Requirements 20° to 22°C
Sufficient For Class of 30 students
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 2
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
S07472ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
152785
Euglena acus Euglena acus N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07473ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
152800
Euglena gracilis Euglena gracilis N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

Low light is 50 to 100 ft. candles of fluorescent light or fluorescent light from the ceiling; High light level is 200 to 400 ft. candles of fluorescent light 18 to 24 inches from the culture.

  • Enough material for a class of 30 students

Specifications

Specifications

Each
Soil Water and Pea
Class of 30 students
Low
20° to 22°C
22°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit