Learn More
Carolina™ Fancy Coleus blumei
A bushy, woody-based evergreen perennial, widely grown for its highly decorative variegated leaves.
Supplier: Carolina Biological Supply 157312
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- Multicolored, variegated clones
- Easy to propagate from stem cuttings
- One plant
Specifications
Widely grown for its highly decorative variegated leaves. | |
Ea. | |
Fancy Coleus |
22°C | |
Coleus blumei |