Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Drosophila Chromosome 2 Mutants
SCI_ED

One of the most widely used model organisms in the world, these flies express a number of eye, wing and other mutations that are easy to see.

Specifications

Quantity Each
For Use With (Application) Students can easily study the sex-linked, recessive and dominant mutations.
Includes Large, vigorous cultures of D. melanogaster in shatterproof 4 x 1.25 in. dia. vials
Media Type Carolina™ Formula 4-24™ Instant Drosophila Medium
Storage Requirements 20° to 25°C
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Includes:

  • Large, vigorous cultures of D. melanogaster in shatterproof 4 x 1.25 in. diameter vials


Products 2
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Scientific Name Price Quantity & Availability  
S07677ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
172460
Chromosome 2 Mutant Drosophila vestigial Drosophila vestigial N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07685ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
172320
Chromosome 2 Mutant Drosophila apterous Drosophila apterous N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

  • Short generation times
  • Sequenced genome
  • Affordable
  • Minimal storage space
  • Truly a model organism

Specifications

Specifications

Each
Large, vigorous cultures of D. melanogaster in shatterproof 4 x 1.25 in. dia. vials
20° to 25°C
Students can easily study the sex-linked, recessive and dominant mutations.
Carolina™ Formula 4-24™ Instant Drosophila Medium
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit