Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Hydra
SCI_ED

An underwater wonder!

Specifications

Quantity Each
Packaging Jar
Storage Requirements 21°C
Sufficient For Class of 30 students
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S07265CND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
TF132800LC
Hydra Coupon N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07265ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
132800
Hydra N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

Generally, the brown hydra species shipped is Hydra littoralis. Specimens are freshly collected to ensure maximum natural size and optimum physiological condition.

  • Easily maintained in a freshwater aquarium
  • For a class of 30 students
  • Needs no filtration or aeration system
  • Reproduces rapidly when fed daily

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

Specifications

Each
Jar
21°C
Class of 30 students
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit