Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Green Algae Cultures
SCI_ED

Green algae are eukaryotic organisms that follow a reproduction cycle called alternation of generations.

$8.35 - $9.10

Specifications

Quantity Each
Includes Carolina™ culturing algae manual
Lighting Type High
Sufficient For Class of 30 students
Temperature (Metric) Range 22°C
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Includes:
  • Materials for a class of 30 students
  • Carolina™ Culturing Algae manual

Products 8
Catalog Number Mfr. No. Product Type Media Type Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Media Type Scientific Name Price Quantity & Availability  
S07464ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152030
Chlamydomonas Soil Water Chlamydomonas
Each for $8.35
Add to cart
 
S07466ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152105
Cladophora Alga-Gro™ seawater Cladophora
Each for $8.35
Add to cart
 
S07465ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152069
Chlorella Alga-Gro™ freshwater Chlorella
Each for $9.10
Add to cart
 
S07483ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152510
Scenedesmus Alga-Gro™ freshwater Scenedesmus
Each for $8.35
Add to cart
 
S07478ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152345
Micrasterias Soil Water Micrasterias
Each for $8.35
Add to cart
 
S07484ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152525
Spirogyra Alga-Gro™ freshwater Spirogyra
Each for $8.35
Add to cart
 
S07487ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152655
Volvox aureus Alga-Gro™ freshwater or soil water Volvox aureus
Each for $8.70
Add to cart
 
S07488ND
SCI_ED
View Documents Promotion Details
Carolina Biological SupplySupplier Diversity Partner
152665
V. globator Alga-Gro™ freshwater or soil water V. globator
Each for $8.70
Add to cart
 
Description

Description

Low light is 50 to 100 ft. candles of fluorescent light or fluorescent light from the ceiling; High light level is 200 to 400 ft. candles of fluorescent light 18 to 24 inches from the culture.

Specifications

Specifications

Each
High
22°C
Carolina™ culturing algae manual
Class of 30 students
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit