Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Sarcodine Cultures
SCI_ED

Sarcodines are single-celled life-forms characterized by an irregular shape.

Specifications

Quantity Each
Includes Dropping pipet, appropriate medium
Packaging Jar
Storage Requirements 20° to 22°C
Sufficient For Class of 30 students
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Includes:
  • Materials for a class of 30 students
  • Dropping pipet
  • Appropriate medium

Products 1
Catalog Number Mfr. No. Product Type Media Type Scientific Name Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Media Type Scientific Name Price Quantity & Availability  
S07217ND
SCI_ED
View Documents
Carolina Biological SupplySupplier Diversity Partner
131306
Amoeba proteus Chilomonas and Wheat Seed with Carolina™ Springwater Amoeba proteus N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

Note: To ensure you order enough living materials for the number of students in your class, order 1 culture for up to 30 students, 2 cultures for 31 to 60 students, 3 cultures for 61 to 90 students, etc.

Specifications

Specifications

Each
Jar
Class of 30 students
Dropping pipet, appropriate medium
20° to 22°C
22°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit