Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina Glove Jersey Gloves Non-distribution product as customer accommodation.

$48.00 - $51.35

Specifications

Color Brown
Material Cotton
For Use With (Application) comfort and breathability in general purpose application
View More Specs

Products 2
Catalog Number Mfr. No. Size Glove Cuff Style Price Quantity & Availability  
Catalog Number Mfr. No. Size Glove Cuff Style Price Quantity & Availability  
19-052996 Non-distribution product as customer accommodation.
View Documents
Carolina Glove
127
Women's Snug knit Pack of 12 for $48.00
Case of 18 PK for $771.48
Add to cart
 
19-059730 Non-distribution product as customer accommodation.
View Documents
Carolina Glove
333
Men's Knit Dozen of 12 for $51.35
Case of 18 DZ for $857.52
Add to cart
 
Specifications

Specifications

Brown
Cotton
comfort and breathability in general purpose application
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit